Please login to your online account to display your discounted pricing

Platinum™ SuperScript™ One-Step RT-PCR System with Platinum Taq DNA Polymerase DFS Item


Convenient 2X format allows further addition of template and primer at any desired concentration

Manufacturer: life technologies  LS10928034

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10928034

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) RT-PCR
Detection Method Gel Electrophoresis
Includes 25μL SuperScript II RT/Platinum Taq DNA Polymerase mix; 1mL 2X reaction mix; 500μL 5mM magnesium sulfate; 1mL 50mM magnesium sulfate
Quantity 25 reactions
Sample Type RNA
Storage Requirements Store at -20deg.C
Quantity 25 reactions