Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ SuperScript™ One-Step RT-PCR System with Platinum Taq DNA Polymerase DFS Item

Convenient 2X format allows further addition of template and primer at any desired concentration

$355.00 - $1,066.00

Specifications

Detection Method Gel Electrophoresis
For Use With (Application) RT-PCR
Sample Type RNA
Storage Requirements Store at -20deg.C
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

10928034

DFS Item
 
life technologies
LS10928034
25μL SuperScript II RT/Platinum Taq DNA Polymerase mix; 1mL 2X reaction mix; 500μL 5mM magnesium sulfate; 1mL 50mM magnesium sulfate 25 reactions Each for $355.00

10928042

DFS Item
 
life technologies
LS10928042
100μL SuperScript II RT/Platinum Taq DNA Polymerase mix; 3 x 1mL 2X reaction mix; 500μL 5mM magnesium sulfate; 1mL 50mM magnesium sulfate 100 reactions Each for $1,066.00
Description & Specifications

Specifications

Detection Method Gel Electrophoresis
For Use With (Application) RT-PCR
Sample Type RNA
Storage Requirements Store at -20deg.C

  • The SuperScript One-Step RT-PCR System with Platinum Taq DNA Polymerase is designed for the convenient, sensitive, and reproducible detection and analysis of RNA molecules by RT-PCR
  • Components for both cDNA synthesis and PCR are combined in a single tube, using gene-specific primers and target RNAs from either total RNA or mRNA
  • Reverse transcription automatically follows PCR cycling without additional steps
  • Using the SuperScript One-Step System The system consists of two major components: RT/Platinum Taq Mix and 2X Reaction Mix
  • The RT/Platinum Taq Mix contains a mixture of SuperScript II Reverse Transcriptase and Platinum Taq DNA Polymerase for optimal cDNA synthesis and PCR amplification
  • SuperScript II RT is a modified version of Moloney Murine Leukemia Virus (M-MLV) RT, engineered to reduce RNase H activity and increase thermal stability
  • Platinum Taq DNA polymerase is Taq DNA polymerase complexed with a proprietary antibody that inhibits polymerase activity at ambient temperatures
  • The antibody is denatured and polymerase activity is restored during the denaturation step in PCR cycling at 94°C
  • This provides an automatic “hot start” in PCR, increasing sensitivity, specificity, and yield
  • The 2X Reaction Mix consists of a proprietary buffer system optimized for reverse transcription and PCR amplification, Mg2+ optimized for universal use, deoxyribonucleotide triphosphates, and stabilizers

  • Final product size: 3kb or less

One-Step RT-PCR, PCR & Real-Time PCR, RT-PCR, Reverse Transcription

Shipping Condition: Wet ice