Please login to your online account to display your discounted pricing

Platinum™ SuperScript™ III One-Step RT-PCR System with Platinum Taq High Fidelity DNA Polymerase DFS Item


Offers highest sensitivity for end point detection

Manufacturer: life technologies  LS12574035

 View more versions of this product

 Disclaimers

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Catalog No. 12574035

  • / Each
 

Description & Specifications

Specifications

Includes 100μL SuperScript III RT/Platinum Taq High Fidelity Enzyme Mix; 3 × 1mL 2X Reaction Mix (containing 0.4mM of each dNTP, 2.4mM MgSO4); 500μL 5mM Magnesium Sulfate
Detection Method Gel Electrophoresis
For Use With (Application) RT-PCR
Sample Type RNA
Storage Requirements Store all components at -30deg. to -10deg.C.
Quantity 100 reactions