missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ CellsDirect™ One-Step qRT-PCR Kit with ROX, for ABI 7000, 7300, 7700, and 7900(HT) instruments DFS Item

RNA purification step not needed, saves time and money

Manufacturer:  Invitrogen™ 11754100

 View more versions of this product

Catalog No. 11754100



Description

Description

The CellsDirect One-Step qRT-PCR Kit delivers highly sensitive and specific real-time qPCR results directly from cells, without the need for an RNA purification step.
Streamline Your Workflow In traditional real-time quantitative RT-PCR (qRT-PCR), you first isolate RNA from cells in a time-consuming procedure that can lead to loss of material. Using the CellsDirect One-Step qRT-PCR Kit, you lyse the cells and add the complete lysate directly to a one-step qRT-PCR reaction with minimal handling and sample loss.
Ideal for Your qRT-PCR Application CellsDirect technology is validated for gene expression studies involving limited samples, and is suitable for validation of RNAi knockdowns. CellsDirect One-Step qRT-PCR Kits are especially valuable for high-throughput applications by eliminating costly, time-consuming RNA purification steps. The CellsDirect One-Step qRT-PCR Kit may also be used to detect genomic DNA targets.
CellsDirect One-Step qRT-PCR Kits may be used in conjunction with hydrolysis probes (e.g., TaqMan™) or LUX™ fluorogenic primers.

  • Efficiency: Offers significant time and cost savings for increased throughput
  • Convenience: Enzyme mix includes both SuperScript™ III Reverse Transcriptase and Platinum™ Taq DNA Polymerase
  • Sensitivity: No separate RNA isolation step increases sensitivity by minimizing sample loss
  • Broad Detection Range: Detect targets from samples ranging from a single cell to 10,000 cells
  • Versatility: Compatible with a wide variety of cell lines and samples collected by Laser Capture Microdissection (LCM)
  • For use with ABI 7000, 7300, 7700, and 7900(HT) instruments

This kit includes 2X Reaction Mix containing ROX™ Reference Dye. An equivalent kit containing a separate tube of ROX Reference Dye, is also available.

Gene Expression Analysis and Genotyping, PCR and Real-Time PCR, Real Time PCR (qPCR), Real Time PCR-Based Gene Expression Profiling, Real-Time PCR Directly from Samples

Specifications

Specifications

2X
Cells
1-Step qRT-PCR, 2-Step qRT-PCR
100
Fast, Standard
Primer-Probe Detection
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, QuantStudio 3 and 5 systems, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 12k Flex system, QuantStudio 6 Flex system
10mL Resuspension Buffer; 1mL Lysis Enhancer; 500μL DNase I, Amplification Grade (1U/μL); 160μL 10X DNase I Buffer; 400μL 25mM EDTA; 100μL SuperScript III RT/Platinum Taq Mix; 2 x 1.25mL 2X Reaction Mix with ROX; 1mL 50mM MgSO4; 2mL DEPC-treated water; 10μL HeLa Total RNA (10ng/μL)
CellsDirect™, Platinum™, SuperScript™
SDS
Documents
Product Certifications
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.