missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ SuperFi™ Green DNA Polymerase DFS Item

Amplify even difficult and GC-rich amplicons with superior accuracy (>100X Taq) and minimaloptimization steps

$254.00 - $4,960.00

Specifications

Storage Requirements Store at -20°C.
Format Separate components
Overhang Blunt
For Use With (Application)
  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates 
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR (up to 20 kb)
  • Fast PCR
Includes Platinum SuperFi DNA Polymerase, 5X SuperFi Green Buffer, 5X SuperFi GC Enhancer
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12357010 Invitrogen™
LS12357010
100 Units Each for $254.00
12357050 Invitrogen™
LS12357050
500 Units Each for $1,166.00
12357250 Invitrogen™
LS12357250
5 x 500 Units Each for $4,960.00
Specifications

Description

Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase which combines superior fidelity with trusted PlatinumTM hot-start technology for the highest success in PCR. Featuring >100x Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis and other applications benefiting from supreme sequence accurac

  • Error-free PCR products due to exceptional >100X Taq fidelity
  • High specificity and increased yields with Platinum hot-start technology
  • Robust amplification of difficult-to-amplify targets including those of sub-optimal purity or with ˃65% GC content 
  • Convenient workflow with room temperature reaction setup and direct gel loading with green buffer format
Description & Specifications

Specifications

Store at -20°C.
Blunt
Platinum SuperFi DNA Polymerase, 5X SuperFi Green Buffer, 5X SuperFi GC Enhancer
Separate components
  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates 
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR (up to 20 kb)
  • Fast PCR