Invitrogen™ Platinum™ SuperFi™ Green DNA Polymerase DFS Item
Manufacturer:  Invitrogen™ 12357002
Catalog No. 12357002
Description
Description
Specifications
Specifications
20 Units
Store at -20C.
  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates 
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR (up to 20 kb)
  • Fast PCR
Blunt
Separate components
Platinum SuperFi DNA Polymerase, 5X SuperFi Green Buffer, 5X SuperFi GC Enhancer