Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ SuperFi™ Green DNA Polymerase DFS Item


Amplify even difficult and GC-rich amplicons with superior accuracy (>100X Taq) and minimaloptimization steps

Manufacturer: life technologies  LS12357050

 View more versions of this product

Catalog No. 12357050

  • / Each
 

Description & Specifications

Specifications

For Use With (Application)
  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates 
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR (up to 20 kb)
  • Fast PCR
Includes Platinum SuperFi DNA Polymerase, 5X SuperFi Green Buffer, 5X SuperFi GC Enhancer
Overhang Blunt
Quantity 500 Units
Format Separate components
Storage Requirements Store at -20C.

Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase which combines superior fidelity with trusted PlatinumTM hot-start technology for the highest success in PCR. Featuring >100x Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis and other applications benefiting from supreme sequence accurac