Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ PCR SuperMix DFS Item

Ready-to-use mixture reduces set up time by 50%

$185.00 - $8,830.00

Specifications

For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR
Overhang 3'-A
Size (Final Product) 5kb or less
View More Specs
Includes:  Platinum PCR SuperMix is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 5000 amplification reactions of 50μL each are provided. Platinum PCR SuperMix Components: 22U/mL complexed recombinant Taq DNA polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM MgCl2, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, and stabilizers. 100 rxns: 4 x 1.125mL
Requires:  Template and primers

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11306081

DFS Item
 
life technologies
LS11306081
5000 reactions Each for $8,830.00

11306016

DFS Item
 
life technologies
LS11306016
100 reactions Each for $185.00
Description & Specifications

Specifications

For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR
Overhang 3'-A
Size (Final Product) 5kb or less

Platinum PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.

  • Allows for room temperature reaction assembly
  • Amplifies fragments up to 5kb
  • Produces products with 3'-A ends
  • Improves PCR specificity
Using Platinum PCR SuperMix
Platinum PCR SuperMix is for high-specificity DNA amplification. High specificity is provided by a mixture of recombinant Taq DNA Polymerase and Platinum monoclonal antibodies to Taq DNA Polymerase. Automatic “hot-start” improves the specificity of PCR. Taq DNA Polymerase is activated during the initial denaturation step.

Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR

Shipping condition: Dry Ice