Please login to your online account to display your discounted pricing

Platinum™ PCR SuperMix DFS Item


Ready-to-use mixture reduces set up time by 50%

Manufacturer: life technologies  LS11306081

 View more versions of this product

Template and primers

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11306081

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR
Includes Platinum PCR SuperMix is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 5000 amplification reactions of 50μL each are provided. Platinum PCR SuperMix Components: 22U/mL complexed recombinant Taq DNA polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM MgCl2, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, and stabilizers. 5000 rxns: 4 x 56.25mL
No. of Reactions 5000.0
Overhang 3'-A
Quantity 5000 reactions
Size (Final Product) 5kb or less