missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ II Taq Hot-Start DNA Polymerase DFS Item

Invitrogen™ Taq Hot-Start DNA Polymerase for fast and easy PCR

Manufacturer:  Invitrogen™ LS14966000

 View more versions of this product

Catalog No. 14966000



Includes: 10 µL Platinum II Taq HS DNA Polymerase, 1.25 mL 5X Platinum II PCR Buffer, 1.25 mL Platinum GC Enhancer

Description

Description

Invitrogen™ Platinum™ Taq II Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq, and leading hot-start technology.

  • Innovative buffer—enables universal primer annealing protocol by isostabilizing primer-template duplex structures
  • Engineered Taq—confers  fast cycling and  tolerance to common inhibitors
  • Platinum hot-start technology—ensures superior specificity, sensitivity, and yields; allows for room temperature reaction setup
  • Optional green buffer—reduces pipetting error with direct gel loading

Additional:

  • Antibody-mediated hot-start
  • Universal annealing protocool
  • Inhibitor resistance

Applications:

  • Genotyping
  • High-throughput PCR
  • GC-rich templates
  • Fast PCR
  • Amplification of samples with suboptimal purity
  • Direct PCR with blood
  • PCR from DNA extracted from FFPE samples
  • Colony PCR
  • PCR from bacterial 16S rRNA genes
Specifications

Specifications

5' to 3'
Genotyping; PCR
GC-rich amplification
Taq HS DNA polymerase; PCR buffer; GC enhancer
25 reactions
Store at -20°C
1X
Separate components
No direct gel loading
3' A
15sec/kb
Target length: Up to 5kb
Documents

For research use only