Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ Taq Antibody DFS Item

A thermolabile inhibitor

$279.00 - $279.00

Specifications

Storage Requirements Store in freezer (-5° to -30°C).
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10965028

DFS Item
 
life technologies
LS10965028
250 units Each for $279.00
Description & Specifications

Specifications

Storage Requirements Store in freezer (-5° to -30°C).

PlatinumTaq Antibody contains monoclonal antibodies to Taq DNA Polymerase for “hot-start” PCR.

  • Alphabetical Ab Target Filter: P,T
  • Unconjugated

PCR, PCR and Real-Time PCR, Routine PCR

Shipping Condition: Approved for shipment on wet or dry ice