Please login to your online account to display your discounted pricing
Platinum™ Pfx DNA Polymerase DFS Item
Built-in hot start
Manufacturer: life technologies  LS11708039
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 11708039
  • / Each
Description & Specifications
Specifications
Fidelity (vs. Taq) Fidelity (vs. Taq): 26X
Gel Compatibility Gel Compatibility: Medium
Includes Platinum Pfx DNA Polymerase (500 units total, at 2.5U/μL) is a supplied with a vial (3 x 1mL) of 10X PCRx Enhancer solution, a vial (3 x 1mL) of 10X Pfx amplification buffer, and a vial (1mL) of 50mM MgSO4.Designed for 500 reactions (at 1 unit each in a 50μL reaction volume).
Quantity 500 reactions
Storage Requirements Store at -20°C.