Platinum™ Pfx DNA Polymerase DFS Item
Built-in hot start
Manufacturer:  Life Technologies LS11708021
 View more versions of this product
Catalog No. 11708021
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Platinum Pfx DNA Polymerase (250 units total, at 2.5U/μL) is a supplied with a vial (2 x 1mL) of 10X PCRx Enhancer solution, a vial (2 x 1mL) of 10X Pfx amplification buffer, and a vial (1mL) of 50mM MgSO4. Designed for 250 reactions (at 1 unit each in a 50μL reaction volume).
Store at -20°C.
Gel Compatibility: Medium
250 reactions
Fidelity (vs. Taq): 26X