Platinum™ GenoType Tsp DNA Polymerase DFS Item
Built-in hot start allows for room temperature reaction
Manufacturer:  Life Technologies LS11448032
 View more versions of this product
Catalog No. 11448032
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Store at -20°C. Guaranteed stable for 6 months when properly stored.
2500 units
1 tube of 500μL (5 Tsp U/μL); 10X PCR Buffer (200mM Tris-HCl pH 8.4, 500mM KCl); 1 vial of 50mM MgCl2
Gel Compatibility: Low
Fidelity (vs. Taq): 1X