Please login to your online account to display your discounted pricing
Platinum™ Blue PCR SuperMix DFS Item
Premixed reagents for high-throughput applications
Manufacturer: life technologies  LS12580023
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 12580023
  • / Each
Description & Specifications
Specifications
For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR
Includes 4 x 11.25mL Composition: Each tube contains 22U/mL complexed recombinant Taq DNA Polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM magnesium chloride, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, stabilizers, glycerol, and blue tracking dye.
Overhang 3'-A
Quantity 1000 reactions
Size (Final Product) 5kb or less