Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ Blue PCR SuperMix DFS Item

Premixed reagents for high-throughput applications

$189.00 - $1,618.00

Specifications

For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR
Overhang 3'-A
Size (Final Product) 5kb or less
View More Specs
Includes:  4 x 1.125mL, Composition: Each tube contains 22U/mL complexed recombinant Taq DNA Polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM magnesium chloride, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, stabilizers, glycerol, and blue tracking dye.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

12580023

DFS Item
 
life technologies
LS12580023
4 x 11.25mL Composition: Each tube contains 22U/mL complexed recombinant Taq DNA Polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM magnesium chloride, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, stabilizers, glycerol, and blue tracking dye. 1000 reactions Each for $1,618.00

12580015

DFS Item
 
life technologies
LS12580015
4 x 1.125mL Composition: Each tube contains 22U/mL complexed recombinant Taq DNA Polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM magnesium chloride, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, stabilizers, glycerol, and blue tracking dye. 100 reactions Each for $189.00
Description & Specifications

Specifications

For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR
Overhang 3'-A
Size (Final Product) 5kb or less

Platinum Blue PCR SuperMix provides qualified reagents for the amplification of nucleic acid templates by PCR, and includes blue tracking dye for subsequent gel analysis. The reagents are premixed for high-throughput applications as well as general ease of use.

Using Platinum Blue PCR SuperMix
The mixture contains recombinant Platinum Taq DNA polymerase, Mg++, dNTPs, glycerol (used as a precipitant), and blue tracking dye. Platinum Blue PCR SuperMix is supplied at a 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents are provided sufficient for 100 or 1,000 amplification reactions of approx. 50μL each.

Hot-start specificity
Specific binding of the anti-Taq antibody inhibits polymerase activity at room temperature. Activity is restored after a denaturation step in PCR cycling at 94°C, providing an automatic “hot start” in PCR and improving specificity and yield.

Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR

Shipping condition: Dry Ice