Please login to your online account to display your discounted pricing

Platinum™ Blue PCR SuperMix DFS Item


Premixed reagents for high-throughput applications

Manufacturer: life technologies  LS12580015

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12580015

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR
Includes 4 x 1.125mL Composition: Each tube contains 22U/mL complexed recombinant Taq DNA Polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM magnesium chloride, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, stabilizers, glycerol, and blue tracking dye.
No. of Reactions 100.0
Overhang 3'-A
Quantity 100 reactions
Size (Final Product) 5kb or less