missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Phasemaker™ Tubes DFS Item

Invitrogen™ Phasemaker™ Tubes contain a polymer that clearly separates and completely isolates the upper aqueous phase of the TRIzol™ mix from the organic phase underneath, making it very easy to pipette off the upper phase containing the RNA.

Supplier:  Invitrogen™ A33248

Catalog No. A33248



Includes: One bag of 100 Phasemaker Tubes

Description

Description

Invitrogen™ Phasemaker™ Tubes contain a polymer that clearly separates and completely isolates the upper aqueous phase of the TRIzol™ mix from the organic phase underneath, making it very easy to pipette off the upper phase containing the RNA. The polymer works on a density gradient, as it is heavier than the aqueous phase, but lighter than the organic phase; after centrifugation it positions itself between these two layers. It creates a tight seal between the two phases, making it very easy to pipette off the aqueous phase without  the risk of carrying over contaminants from the organic layer. Because this, Invitrogen Phasemaker Tubes allow recovery of the whole aqueous phase, increasing yield up to 30%. It is an inert polymer that does not interfere with downstream experiments such as PCR. It works for RNA extraction using TRIzol™, TRIzol™ LS and TRIzol™ Plus only. It is not recommended to be used with other organic reagents as their density might be different. It works with fresh or frozen samples (tissue or blood). It also  works with tissue samples treated previously with RNAlater. Invitrogen Phasemaker Tubes come already pre-dispensed. The tube size is  2 mL.

  • Pre-dispensed in 2 mL tubes
  • Creates a tight seal between the aqueous and the organic phases
  • Easier to pipette off the whole aqueous phase
  • Increases yield up to 30%
  • RNA integrity is not compromised
  • The seal stays for longer periods of time, allowing you to increase the number of samples per batch
  • Inert, does not interfere with downstream experiments

Warranty

One year after receiving

Specifications

Specifications

100 Phasemaker Tubes; room temperature

Total RNA
100 Tubes
Cells: ≤107
Tissue: ≤100 mg
Cells, Tissue
Not High-throughput Compatible (Manual)
Room Temperature
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.