Please login to your online account to display your discounted pricing

PCR SuperMix DFS Item


Ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification

Manufacturer: life technologies  LS10572063

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10572063

  • / Each
 

Description & Specifications

Specifications

Exonuclease Activity 5' - 3'
Fidelity 1 X
For Use With (Application) Efficient PCR amplifications
GC-Rich PCR Performance Low
Includes Contains 4 bottles (56.25mL each) of PCR SuperMix. Components: 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM Magnesium Chloride, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, 22U/mL recombinant Taq DNA Polymerase, and stabilizers
No. of Reactions 5000.0
Polymerase Taq DNA Polymerase
Product Type PCR SuperMix
Format SuperMix
Overhang 3'-A
Quantity 5000 reactions
Size (Final Product) 5 kb or less