missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PCR SuperMix DFS Item

Ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification

$129.00 - $7,129.50

Specifications

Exonuclease Activity 5' - 3'
Fidelity 1 X
Format SuperMix
For Use With (Application) Efficient PCR amplifications
GC-Rich PCR Performance Low
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Contains four vials (1.125mL each) of PCR SuperMix. Components: 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM Magnesium Chloride, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, 22U/mL recombinant Taq DNA Polymerase, and stabilizers.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10572063 Life Technologies
LS10572063
5000 reactions Each for $7,129.50
10572014 Life Technologies
LS10572014
100 reactions Each for $129.00
Specifications

Description

Simply add template and primers, reducing set-up time by half. PCR SuperMix contains Mg2+, dNTPs, and recombinant Taq DNA Polymerase at concentrations sufficient for routine PCR of fragments up to 5kb. PCR SuperMix is supplied at a 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions.

ChIP-on-Chip, Chromatin Biology, PCR, PCR and Real-Time PCR, RNAi, Epigenetics and Non-Coding RNA Research, Routine PCR

Order Info

Shipping Condition: Approved for shipment on wet or dry ice

Description & Specifications

Specifications

5' - 3'
SuperMix
Low
PCR SuperMix
1 X
Efficient PCR amplifications
Taq DNA Polymerase