missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PCR Optimized Buffers DFS Item

Use with PCR Optimizer Kit to provide optimal buffer conditions for all primer-template combinations

Manufacturer:  Invitrogen™ LSK122002J

 View more versions of this product

Catalog No. K122002J


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications Description & Specifications

Specifications

Buffer J
Store at -20°C. All components are guaranteed stable for 6 months when properly stored.
The PCR Optimizer™ Kit contains 10 5X reaction buffers (which vary in pH and magnesium concentrations), sterile water, DMSO, and preneutralized dNTPs. The Optimized™ Buffer Kits contain the Optimized™ Buffer of your choice (A-J), preneutralized dNTPs, sterile water, and DMSO. dNTPs are also available separately.
Taq DNA Polymerase
200.0