Please login to your online account to display your discounted pricing

Invitrogen™ PCR Optimized Buffers DFS Item


Use with PCR Optimizer Kit to provide optimal buffer conditions for all primer-template combinations

Manufacturer: invitrogen™  LSK122002I

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. K122002I

  • / Each
 

Description & Specifications

Specifications

Includes The PCR Optimizer™ Kit contains 10 5X reaction buffers (which vary in pH and magnesium concentrations), sterile water, DMSO, and preneutralized dNTPs. The Optimized™ Buffer Kits contain the Optimized™ Buffer of your choice (A-J), preneutralized dNTPs, sterile water, and DMSO. dNTPs are also available separately.
No. of Reactions 200.0
Polymerase Taq DNA Polymerase
Product Type Buffer I
Storage Requirements Store at -20°C. All components are guaranteed stable for 6 months when properly stored.