missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PCR Optimized Buffers DFS Item

Use with PCR Optimizer Kit to provide optimal buffer conditions for all primer-template combinations

$394.80 - $400.05

Specifications

Includes The PCR Optimizer™ Kit contains 10 5X reaction buffers (which vary in pH and magnesium concentrations), sterile water, DMSO, and preneutralized dNTPs. The Optimized™ Buffer Kits contain the Optimized™ Buffer of your choice (A-J), preneutralized dNTPs, sterile water, and DMSO. dNTPs are also available separately.
No. of Reactions 200.0
Storage Requirements Store at -20°C. All components are guaranteed stable for 6 months when properly stored.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: The PCR Optimizer™ Kit contains 10 5X reaction buffers (which vary in pH and magnesium concentrations), sterile water, DMSO, and preneutralized dNTPs. The Optimized™ Buffer Kits contain the Optimized™ Buffer of your choice (A-J), preneutralized dNTPs, sterile water, and DMSO. dNTPs are also available separately.


Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
K122002A Invitrogen™
LSK122002A
Buffer A Each for $399.00
K122002B Invitrogen™
LSK122002B
Buffer B Each for $399.00
K122002C Invitrogen™
LSK122002C
Buffer C Each for $396.90
K122002D Invitrogen™
LSK122002D
Buffer D Each for $399.00
K122002H Invitrogen™
LSK122002H
Buffer H Each for $394.80
K122002F Invitrogen™
LSK122002F
Buffer F Each for $400.05
K122002E Invitrogen™
LSK122002E
Buffer E Each for $396.90
K122002I Invitrogen™
LSK122002I
Buffer I Each for $394.80
K122002G Invitrogen™
LSK122002G
Buffer G Each for $394.80
K122002J Invitrogen™
LSK122002J
Buffer J Each for $396.90
Specifications

Description

Optimized™ Buffer Kits available in convenient 200- and 800-reaction sizes to ensure perfect PCR every time. Varying pH level, magnesium concentration, and DMSO content in PCR alters specificity of priming and eliminates background products. This enhances the performance of all primer-template combinations, even those with limited homology. Once optimal PCR conditions have been identified, use these individual PCR Optimizer™ Buffers and dNTPs to provide optimal buffer conditions for all primer-template combinations. Each buffer provides 200 reactions.

  • Available in convenient 200- and 800-reaction sizes to ensure perfect PCR

PCR, PCR and Real-Time PCR, Routine PCR

Order Info

Shipping Condition: Dry ice

Description & Specifications

Specifications

The PCR Optimizer™ Kit contains 10 5X reaction buffers (which vary in pH and magnesium concentrations), sterile water, DMSO, and preneutralized dNTPs. The Optimized™ Buffer Kits contain the Optimized™ Buffer of your choice (A-J), preneutralized dNTPs, sterile water, and DMSO. dNTPs are also available separately.
200.0
Store at -20°C. All components are guaranteed stable for 6 months when properly stored.