Please login to your online account to display your discounted pricing

One Shot™ TOP10 Chemically Competent E. coli, High Copy Plasmid DFS Item


Ideal for high-efficiency cloning and plasmid propagation

Manufacturer: life technologies  LSC404010

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. C404010

  • / Each
 

Description & Specifications

Specifications

Cell Type Chemically Competent
Contains One Shot TOP10 Chemically Competent E. coli: 11 vials, 50μL each, pUC19 DNA (10pg/μL): 1 vial, 50μL, S.O.C. medium: 1 bottle, 6mL
Contains F' Episome No
Recombinant Yes
Species E. coli
Quantity 10 reactions
Storage Requirements Store Competent Cells at -80°C. Store pUC19 DNA at -20°C. Store SOC Medium at 4°C or at room temperature.
Efficiency > 1x109
T1 Phage Resistant No
Includes Blue/White Screening Yes
Improves Plasmid Quality Yes
Bacterial Content TOP10
For Use With (Application) Cloning, Transformation