Please login to your online account to display your discounted pricing

MultiShot™ StripWell TOP10 Chemically Competent E. coli DFS Item


Allows rapid transformation in a convenient 96-well format

Manufacturer: life technologies  LSC409601

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. C409601

  • / Each
 

Description & Specifications

Specifications

Cell Type Chemically Competent
Bacterial Content TOP10
Contains MultiShot StripWell TOP10 Chemically Competent E. coli: One 96-stripwell plate, 50μL per well, pUC19 DNA (10pg/μL): 1 vial, 50μL, S.O.C. medium: 2 bottles, 15mL each
Contains F' Episome No
Efficiency > 1x108
T1 Phage Resistant No
Improves Plasmid Quality Yes
Includes Blue/White Screening Yes
Recombinant Yes
Species E. coli
For Use With (Application) Cloning, Transformation
Quantity 1 plate
Storage Requirements Store Competent Cells at -80°C. Store pUC19 DNA at -20°C. Store SOC Medium at 4°C or at room temperature.