missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Mach1™ T1 Phage-Resistant Chemically Competent E. coli DFS Item

Clearly visible within eight hours of plating the transformation mix

$560.00 - $3,045.00

Specifications

Bacterial Content Mach1
Cell Type Chemically Competent
Species E. coli
Storage Requirements Store Competent Cells at -80°C. Store pUC19 DNA at -20°C. Store SOC Medium at 4°C or at room temperature.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C862003 Life Technologies
LSC862003
20 x 50μL Each for $560.00
C869601 Life Technologies
LSC869601
1 plate Each for $3,045.00
Specifications

Description

Mach1 T1 Phage-Resistant Chemically Competent E. coli are the fastest growing chemically competent cells currently available. Doubling time is approximately 50 minutes, compared to >74 minutes for other cloning strains. Mach1 colonies enabling you to plate and pick colonies the same day. Perform mini-preps after only four hours of growth from an overnight colony—saving an entire day in your experiments.

  • Minimizes nonspecific recombination in cloned DNA (recA)
  • Offers resistance to T1 and T5 phage (tonA)
  • Yields cleaner preparations of DNA and better results in downstream applications due to the elimination of nonspecific digestion by Endonuclease I (endA1)
  • Provides efficient transformation of unmethylated DNA from PCR amplifications (hsdR)
  • Allows blue/white screening of recombinant clones (lacZΔM15)
  • Two formats to choose: Convenient One Shot format provides single-use, 50μL aliquots that eliminate efficiency-zapping, freeze-thaw cycles and money wasted on unused cells
  • MultiShot StripWell format allows rapid, high-throughput transformation in a 96-well stripwell plate, which allows as few or as many transformations as needed without wasting reagents

Not high-throughput compatible (Manual)

Cloning, Transformation

Order Info

Shipping Condition: Dry Ice

Description & Specifications

Specifications

Mach1
Chemically Competent
E. coli
Store Competent Cells at -80°C. Store pUC19 DNA at -20°C. Store SOC Medium at 4°C or at room temperature.