missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Tfi DNA Polymerase DFS Item

A new non-Taq based, PCR enzyme derived from Thermus filiformis for use in PCR applications without any licensing fees

$35.09 - $932.40

Specifications

Storage Requirements Store at -20°C. Guaranteed stable for 6 months when properly stored.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Tfi DNA Polymerase is supplied with a vial of magnesium-free 5X Tfi PCR Buffer and a vial of 50mM MgCl2.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
30342011 Life Technologies
LS30342011
100 Reactions Each for $35.09
30342024 Life Technologies
LS30342024
500 Reactions Each for $171.15
30342052 Life Technologies
LS30342052
5000 Reactions Each for $932.40
Specifications

Description

Like Taq, Tfi has 5' to 3' exonuclease and 5' to 3' polymerase activity. PCR performance is comparable to Taq DNA polymerase in yield, specificity, fidelity, and robustness. Tfi can be substituted for Taq DNA Polymerase in virtually any application and is ideal for genomic, cDNA, and plasmid targets up to 4kb in length. Tfican be considered for use as a component in a diagnostic application if qualified by the end user.

  • Eliminate licensing fees and restrictions associated with Taq-based enzymes
  • Save money-Tfiis less expensive than TaqDNA Polymerase
  • Achieve yield, specificity, and fidelity comparable to Taq DNA polymerase
  • No need to reoptimize - swap out Taq for Tfi

Tfi is also available in special pack sizes and for OEM customers who may require PCR kit manufacturing flexibility.

PCR, PCR and Real-Time PCR, Routine PCR

Order Info

Shipping Condition: Dry ice

Description & Specifications

Specifications

Store at -20°C. Guaranteed stable for 6 months when properly stored.