Please login to your online account to display your discounted pricing
Pfx50™ DNA Polymerase DFS Item
Designer enzyme ideal for highest fidelity and initial PCR success
Manufacturer: life technologies  LS12355036
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 12355036
  • / Each
Description & Specifications
Specifications
Fidelity (vs. Taq) Fidelity (vs. Taq): 50X
Includes Pfx50 DNA Polymerase is supplied with a vial of 10X Pfx50 PCR amplification buffer contains 1.2mM MgSO4 and a vial of 50mM MgSO4 for further optimization.
Gel Compatibility Gel Compatibility: Low
Quantity 500 reactions
Storage Requirements Store at -20°C. Guaranteed stable for 6 months when properly stored.