missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Pfx50™ DNA Polymerase DFS Item

Designer enzyme ideal for highest fidelity and initial PCR success

$879.90 - $879.90

Specifications

Fidelity (vs. Taq) Fidelity (vs. Taq): 50X
Gel Compatibility Gel Compatibility: Low
Storage Requirements Store at -20°C. Guaranteed stable for 6 months when properly stored.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Pfx50 DNA Polymerase is supplied with a vial of 10X Pfx50PCR amplification buffer contains 1.2mM MgSO4 and a vial of 50mM MgSO4 for further optimization.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12355036 Life Technologies
LS12355036
500 reactions Each for $879.90
Specifications

Description

Pfx50™ DNA Polymerase is a designer enzyme derived from the archaean Thermococcus zilligii. In addition to 3' to 5' proofreading ability, Pfx50 polymerase is fused to an accessory protein for enhanced fidelity and processivity. This unique combination makes the Pfx50 enzyme ideal for applications that require the highest fidelity and PCR success the first time.

  • Achieve ultra low error rates—50 times better than Taq, without compromising PCR robustness
  • Save time and money—highest fidelity means you'll sequence fewer clones to obtain error-free constructs
  • Produce results quickly and easily—robust performance with extremely short extension holds
  • Fidelity (vs. Taq): 50X

Downstream compatibility with popular cloning techniques
Pfx50 DNA Polymerase produces blunt-ended PCR products for compatibility with Directional TOPO™ Expression Kits and Zero Blunt™ TOPO PCR Cloning Kits.

Unit Definition:
One unit of Pfx50DNA Polymerase incorporates 10 nmol of deoxyribonucleotide into acid-insoluble material in 30 minutes at 74°C.

Amplification of Bisulfite-Treated DNA, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), High Fidelity PCR, Methylation Analysis, PCR, PCR and Real-Time PCR, RNAi, Epigenetics and Non-Coding RNA Research

Order Info

Shipping Condition: Dry Ice

Description & Specifications

Specifications

Fidelity (vs. Taq): 50X
Gel Compatibility: Low
Store at -20°C. Guaranteed stable for 6 months when properly stored.