Thermo Scientific™ Invitrogen™ EVOS™ FL Auto 2 Imaging System
Designed with advanced capabilities to simplify demanding cell-based imaging applications such as live-cell imaging, image tiling, and Z-stacking. Bring high performance right to your lab bench with the new Invitrogen™ EVOS™ FL Auto 2 Imaging System.
Manufacturer:  Thermo Scientific™ AMAFD2000
Catalog No. AMAFD2000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Speed: scan a 96-well plate in 3 fluorescent channels in less than 5 minutes
  • Flexibility: customize the system with more than 20 user-changeable LED light cubes, dual cameras (monochrome and color), a variety of objectives ranging from 1.25x to 100x, and multiple vessel holders
  • Time-lapse live-cell imaging: option for precise control of temperature, humidity, and gases for normoxic or hypoxic conditions allows a wide range of biological studies under physiological conditions with an onstage incubator
  • Area view: move rapidly and seamlessly between low-magnification, single-field mode and high-magnification scan mode to easily define and capture the area of interest
  • Automation: time-saving features such as autofocus, rapid stage movement, and automated routines help reduce time to complete experiments, allowing high throughput, high data quality, and improved experimental reproducibility
  • Data analysis: optional advanced software package for quantitative and statistical analysis
Specifications
Specifications
Fluorescence microscopy, Live Cell Imaging
EVOS Light Cubes, EVOS Onstage Incubator