missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Elongase™ Enzyme Mix DFS Item

Amplifies both long and short DNA templates

$242.00 - $1,062.00

Specifications

Composition Elongase Enzyme Mix is supplied with 5X buffer A and 5X buffer B. PCR SuperMix High Fidelity is supplied in a 1.1X concentration. It contains Elongase Enzyme Mix, dNTPs, salts, and MgSO4.
Fidelity (vs. Taq) Fidelity (vs. Taq): 5X
Gel Compatibility Gel Compatibility: Low
Storage Requirements Store at -20°C. Guaranteed stable for 6 months when properly stored.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10480010 Life Technologies
LS10480010
100 reactions Each for $242.00
10480028 Life Technologies
LS10480028
500 reactions Each for $1,062.00
Specifications

Description

The Elongase Amplification System is ideal for amplifying DNA fragments up to 30 kb from purified DNA or crude samples. The enzyme mix is an optimized mixture of recombinant thermostable DNA polymerases, TaqDNA polymerase, and the proofreading (3'-5' exonuclease activity) enzyme Pyrococcus species GB-Dpolymerase.

  • Five-times higher fidelity than Taq DNA polymerase alone
  • A buffer system that allows amplification of simple and complex genomic targets
  • Amplification of long PCR templates or cDNA templates in RT-PCR applications

Long Fragment PCR, PCR, PCR and Real-Time PCR

Order Info

Shipping Condition: Dry Ice

Description & Specifications

Specifications

Elongase Enzyme Mix is supplied with 5X buffer A and 5X buffer B. PCR SuperMix High Fidelity is supplied in a 1.1X concentration. It contains Elongase Enzyme Mix, dNTPs, salts, and MgSO4.
Fidelity (vs. Taq): 5X
Gel Compatibility: Low
Store at -20°C. Guaranteed stable for 6 months when properly stored.