Elongase™ Enzyme Mix DFS Item
Amplifies both long and short DNA templates
Manufacturer:  Life Technologies LS10480010
 View more versions of this product
Catalog No. 10480010
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Elongase Enzyme Mix is supplied with 5X buffer A and 5X buffer B. PCR SuperMix High Fidelity is supplied in a 1.1X concentration. It contains Elongase Enzyme Mix, dNTPs, salts, and MgSO4.
Gel Compatibility: Low
Store at -20°C. Guaranteed stable for 6 months when properly stored.
Fidelity (vs. Taq): 5X
100 reactions