missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Electroporation Cuvettes, 0.1 cm DFS Item

DNA can be introduced into bacterial, yeast, and mammalian cells by electroporation

$318.00 - $463.00

Specifications

Quantity 50/bag
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
P45050 Invitrogen™
LSP45050
0.2cm Each for $328.00
P41050 Invitrogen™
LSP41050
0.1cm Each for $318.00
P46050 Invitrogen™
LSP46050
0.4cm Each for $463.00
Specifications

Description

  • Electroporation cuvettes for use with mammalian, bacterial or yeast cells that feature: Compatible with virtually all common electroporation devices
  • Gamma-irradiated and individually packaged to ensure sterility
  • Color-coded snap cap for easy identification of gap size
  • Recommended for all electroporation competent cells except catalog #11319-019

Cloning, Transformation

Order Info

Shipping Condition: Room Temperature

Description & Specifications

Specifications

50/bag