Invitrogen™ Electrocomp™ TOP10F Electrocomp Kit DFS Item
Identical to TOP10 with the addition of an F' episome
$1,152.00 - $2,370.00
Specifications
Includes Contains enough electrocompetent cells for 75 x 40μL rxns or 50 x 20μL rxns
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Includes: This kit contains enough electrocompetent cells for 20 x 40μL rxns or 40 x 20μL rxns. Please see the product manual for reaction scale recommendations.
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C66511 Life Technologies
LSC66511
10 x 100 μL Each for $1,152.00
C66524 Life Technologies
LSC66524
30 x 100 μL Each for $2,370.00
Specifications
Description
  • Bacterial/Yeast Strain: TOP10F
Cloning, Transformation
Description & Specifications
Specifications
Contains enough electrocompetent cells for 75 x 40μL rxns or 50 x 20μL rxns