Invitrogen™ Electrocomp™ TOP10F Electrocomp Kit DFS Item
Identical to TOP10 with the addition of an F' episome
Supplier:  Invitrogen™ C66524
Catalog No. C66524
Description
Description
  • Bacterial/Yeast Strain: TOP10F
Cloning, Transformation
Specifications
Specifications
Contains enough electrocompetent cells for 75 x 40μL rxns or 50 x 20μL rxns
30 x 100 μL