missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ E-Gel™ Go! Agarose Gels, 2%
For performing agarose gel electrophoresis with only a few samples to analyze
Manufacturer: Invitrogen™ G441002
Description
- Precast agarose gel cassettes each have four wells and are premixed with a blue light excitable nucleic acid stain
- Can be stored at room temperature and are ready to use without additional buffers or reagents
- Designed specifically to work with the components of the E-Gel Go! System
- For a quick check of end point PCRs, restriction digests, plasmid preps or other molecular biology reactions
- Use proprietary stain formulated to provide the high detection sensitivity and can be visualized with traditional UV transilluminators
- When using blue light the bands must be viewed through an amber filter
- Sharp bands and repeatable, reliable results
- Single comb well format
- Sample Volume; 10μL
- For linear double stranded DNA fragments ranging from 1.5kbp to 100bp in size the 2% gels offer excellent resolution and separation
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
E-Gel™ 1 Kb Plus Express Ladder | |
E-Gel | |
10 µl | |
Proprietary Stain | |
E-Gel™ Go! | |
Room Temperature | |
Single Comb |
2% | |
Go Agarose Gel | |
4-well | |
Each pack contains 10 individually sealed pouches, each with a single 1% E-Gel Go! precast agarose gel cassette. | |
10 gel(s) | |
E-Gel™ Go! |
For Research Use Only. Not for use in diagnostic procedures.