Please login to your online account to display your discounted pricing

Invitrogen™ E-Gel™ Go! Agarose Gels, 2% DFS Item

For performing agarose gel electrophoresis with only a few samples to analyze

$93.00 - $151.00

Specifications

Concentration 2%
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Kit Contents Quantity Price Quantity    

G441002

DFS Item
 
invitrogen™
LSG441002
Each pack contains 10 individually sealed pouches, each with a single 1% E-Gel Go! precast agarose gel cassette. 10 Gels Each for $93.00

G442002

DFS Item
 
invitrogen™
LSG442002
Each pack contains 20 individually sealed pouches, each with a single 1% E-Gel Go! precast agarose gel cassette 20 Gels Each for $151.00
Description & Specifications

Specifications

Concentration 2%

  • Precast agarose gel cassettes each have four wells and are premixed with a blue light excitable nucleic acid stain
  • Can be stored at room temperature and are ready to use without additional buffers or reagents
  • Designed specifically to work with the components of the E-Gel Go! System
  • For a quick check of end point PCRs, restriction digests, plasmid preps or other molecular biology reactions
  • Use proprietary stain formulated to provide the high detection sensitivity and can be visualized with traditional UV transilluminators
  • When using blue light the bands must be viewed through an amber filter
  • Sharp bands and repeatable, reliable results
  • Single comb well format
  • Sample Volume; 10μL

  • For linear double stranded DNA fragments ranging from 1.5kbp to 100bp in size the 2% gels offer excellent resolution and separation

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Shipping Condition: Room Temperature