Invitrogen™ E-Gel™ Go! Agarose Gels, 1% DFS Item
For performing agarose gel electrophoresis with only a few samples to analyze
Manufacturer:  Invitrogen™ G441001
 View more versions of this product
Catalog No. G441001
Description
Description
  • Precast agarose gel cassettes each have four wells and are premixed with a blue light excitable nucleic acid stain
  • Can be stored at room temperature and are ready to use without additional buffers or reagents
  • Designed specifically to work with the components of the E-Gel Go! System
  • For a quick check of end point PCRs, restriction digests, plasmid preps or other molecular biology reactions
  • Use proprietary stain formulated to provide the high detection sensitivity and can be visualized with traditional UV transilluminators
  • When using blue light the bands must be viewed through an amber filter
  • Sharp bands and repeatable, reliable results
  • Single comb well formatSample Volume; 10μL
  • For DNA fragments up to 4kbp
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Specifications
E-Gel™ 1 Kb Plus Express Ladder
1%
Go Agarose Gel
100 bp to 4 kb
4-well
Room Temperature
Single Comb
• Each pack contains 10 individually sealed 1% E-Gel™ Go! precast agarose gel cassettes
Store at room temperature.
E-Gel
10 µL
SYBR™ Gold
10 Gels/Pk.
E-Gel™ Go!
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.