missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ E-Gel™ Go! Agarose Gels, 1%
For performing agarose gel electrophoresis with only a few samples to analyze
$194.00 - $307.00
Specifications
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder |
---|---|
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type Specs | Go Agarose Gel |
Sample Loading Volume | 10 µL |
Description
- Precast agarose gel cassettes each have four wells and are premixed with a blue light excitable nucleic acid stain
- Can be stored at room temperature and are ready to use without additional buffers or reagents
- Designed specifically to work with the components of the E-Gel Go! System
- For a quick check of end point PCRs, restriction digests, plasmid preps or other molecular biology reactions
- Use proprietary stain formulated to provide the high detection sensitivity and can be visualized with traditional UV transilluminators
- When using blue light the bands must be viewed through an amber filter
- Sharp bands and repeatable, reliable results
- Single comb well formatSample Volume; 10μL
- For DNA fragments up to 4kbp
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
E-Gel™ 1 Kb Plus Express Ladder | |
E-Gel | |
10 µL | |
SYBR™ Gold | |
Room Temperature | |
Single Comb |
1% | |
Go Agarose Gel | |
100 bp to 4 kb | |
4-well | |
E-Gel™ Go! |
For Research Use Only. Not for use in diagnostic procedures.