missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ E-Gel™ EX Agarose Gels, 1% DFS Item

Precast, fast-resolving gels with 11 wells and an openable format

Manufacturer:  Invitrogen™ LSG402001

 View more versions of this product

Catalog No. G402001



Requires : Require the E-Gel iBase™ Power System for running and the E-Gel Safe Imager™ Real-Time Transilluminator for real-time viewing of sample migration.

Description

Description

  • Contain a proprietary fluorescent nucleic acid stain (excitation: 490nm, emission: 520nm) that can be viewed with blue light, which minimizes DNA damage and allows detection down to 1ng/band of DNA
  • Complete separation (by molecular weight) in 10 minutes
  • Live monitoring of DNA migration without the hazards of UV light
  • Easy access to the gel for extraction of DNA bands or transfer to membranes for Southern blotting
  • Demonstrate over five-fold greater sensitivity than comparable gels containing ethidium bromide allows less sample use per run
  • Fast analysis of RNA samples to check on their integrity before proceeding with downstream application
  • The gel cassette is designed to open with a Gel Knife, so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis
  • Single comb well format
  • Sample Type: Double-Stranded DNA (dsDNA), Single-Stranded RNA
  • Sample Volume; 20μL

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Specifications

Specifications

Proprietary Stain
E-Gel™
20 µL
Room Temperature
E-Gel EX 20-Paks contain 20 gels.
1 %
11 wells
E-Gel® EX 20-Paks contain 20 gels. Store gels at room temperature. E-Gel® EX gels are guaranteed stable for 3 months when properly stored.
Molecular Weight
Preparative without dyes, Non-preparative
Double-Stranded DNA (dsDNA), Single-Stranded RNA
Single Comb
100 bp to 5 kb
E-Gel
20 Pack
Documents

For Research Use Only. Not for use in diagnostic procedures.