Invitrogen™ E-Gel™ Agarose Gels with SYBR™ Safe DNA Gel Stain, 1.2% Greener Choice DFS Item
Less hazardous
Manufacturer:  Invitrogen™ LSG521801
Catalog No. G521801
Includes: Contains one multi-pack of 18 gels.
Description
Description
  • Ready for use with agarose, electrodes, and DNA stain packaged inside a dry, disposable, UV-transparent cassette
  • There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble
  • Offer excellent resolution and clarity in 12 minutes and are excellent for analyzing PCR products, restriction digests, and plasmid preparations
  • 12-well, single comb gels
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
1.2%
12-well
Room Temperature
E-Gel™
Agarose Gel
E-Gel
SYBR Safe
Single Comb
Less hazardous
SDS
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.