Please login to your online account to display your discounted pricing

Invitrogen™ DYNAL™ Dynabeads™ Pan Mouse IgG, Monoclonal DFS Item

Binds monoclonal mouse IgGs of choice for specific cell isolation or depletion

$802.00 - $3,120.00

Specifications

Applications Cell Analysis, Cell Isolation, Cell Isolation and Expansion, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Human Cell Isolation, RNAi, Epigenetics and Non-Coding RNA Research, Secondary Cell Isolation
Cross Reactivity All species, except mouse
Dye Type Dynabeads™ Pan
No. of Cells 1x107 PBMCs per isolation
Outputs >95%
View More Specs
Includes:  5mL Dynabeads Pan Mouse IgG coated with monoclonal human anti-mouse IgG. The monoclonal antibody is unique Fc-specific human anti-mouse IgG4, and it shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11041

DFS Item
 
life technologies
LS11041
5mL Each for $802.00

11042

DFS Item
 
life technologies
LS11042
5 x 5mL Each for $3,120.00
Description & Specifications

Specifications

Applications Cell Analysis, Cell Isolation, Cell Isolation and Expansion, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Human Cell Isolation, RNAi, Epigenetics and Non-Coding RNA Research, Secondary Cell Isolation
Cross Reactivity All species, except mouse
Dye Type Dynabeads™ Pan
No. of Cells 1x107 PBMCs per isolation
Outputs >95%
Purification Method Depletion, negative isolation or positive isolation
Purity Research Grade
Storage Requirements Storage: 2°C to 8°C.

Dynabeads Pan Mouse IgG will bind all subclasses of mouse IgG and is Fc-specific. Low amount of IgG is required for binding. All cells from all species except from mouse can be isolated with product, depending only on specific antibody of choice.

  • Cells can be depleted from sample
  • Cells can be positively isolated from sample for molecular applications
  • Cells can be negatively isolated from PBMC sample by adding antibody cocktail of mouse IgGs towards unwanted cells
  • Compatible with flow cytometry
  • Starting samples: Whole blood, PBMC, buffy coat, tissue digests
  • Whole Antibody Capacity: Processes approx. 2x109 cells total
  • Cell Type: All cells from all species, except mouse
  • Output Viability: >95%
  • Reactivity: All species, except mouse
  • Sample Type: Blood, PBMC, Tissue Digests
  • Starting Material (Cell No.): 1x107 PBMCs per isolation
  • Target Species: All species, except mouse

For cellular assays we recommend positive isolation followed by release of beads from the cells. Recommended product for bead-free cells: Dynal CELLection Pan Mouse IgG in combination with your mouse IgG.

Cell Analysis, Cell Isolation, Cell Isolation and Expansion, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Human Cell Isolation, RNAi, Epigenetics and Non-Coding RNA Research, Secondary Cell Isolation

Shipping Condition: Room Temperature