Please login to your online account to display your discounted pricing

DYNAL™ Dynabeads™ M-280 Streptavidin DFS Item


For solation and handling of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets

Manufacturer: life technologies  LS11205D

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11205D

  • / Each
 

Description & Specifications

Specifications

Diameter (Metric) Particle 2.8μm
Includes Dynabeads (2.8μm) with covalently coupled recombinant streptavidin. Supplied in PBS, pH 7.4 with 0.1% BSA and 0.02% sodium azide added as a preservative
Applications Antibody Purification, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), DNA and RNA Purification and Analysis, DNA Extraction, Immunoprecipitation, Phage Display, Protein and Peptide Purification, Protein Complex Isolation and Analysis, Protein Complex Pull-Down, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis, RNAi, Epigenetics and Non-Coding RNA Research, Sanger Sequencing, Sequencing, Sequencing Reaction Purification
Purity Research Grade
Quantity 2mL
Sample Type Any Sample Type
Shelf Life 36 months
Storage Requirements Storage: 2°C to 8°C.
Concentration 10mg/mL