missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynabeads™ M-270 Amine
These are hydrophilic Dynabeads™ (2.8 μm) with amine groups. The surface amino groups allow direct covalent binding of carbohydrates, glycoproteins and glycolipids through reductive amination of aldehydes.
$808.00 - $2675.00
Specifications
| Content And Storage | Store in refrigerator (2–8°C). |
|---|---|
| Diameter (Metric) | 2.8 μm |
| Elution Volume Min | 5 μL |
| High-throughput Compatibility | High-throughput Compatible |
| Shipping Condition | Room Temperature |
Description
The surface amino groups allow direct covalent binding of carbohydrates, glycoproteins and glycolipids through reductive amination of aldehydes. For use with any sample type.
- Rapid binding (less than 1 hour) at neutral to high pH and room temperature
- No further surface activation required
- Excellent reactivity with a range of cross-linking reagents, allowing for easy introduction of further alternative surface chemistries
- Ideal for isolation of small components such as proteins and peptides
- Allows C-terminal coupling of proteins
- Well suited for isolation of proteins or other targets with affinity for specific carbohydrate moieties
Order Info
Shipping Condition: Room Temperature
Specifications
| Store in refrigerator (2–8°C). | |
| 5 μL | |
| Room Temperature | |
| Amine | |
| Bead with Amine Group | |
| Research Grade |
| 2.8 μm | |
| High-throughput Compatible | |
| Any Sample Type | |
| Dynabeads | |
| Beads in suspension |
For Research Use Only. Not for use in diagnostic procedures.