Please login to your online account to display your discounted pricing

dNTP Set (100 mM) Solution DFS Item


Ready-to-use solution

Manufacturer: life technologies  LS10297018

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10297018

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Includes 100mM each of dATP, dCTP, dGTP, and dTTP; Each vial contains 250μL (25μmol) dNTP in purified water
Concentration 100mM
Storage Requirements Store at -30° to -10°C.
Quantity 4 x 250μL