Please login to your online account to display your discounted pricing
DNaseAlert™ QC System DFS Item
Provides sensitive detection of DNases in a simple, easy-to-use fluorimetric assay with real time results
Manufacturer: life technologies  LSAM1970M
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1970M
  • / Each
Description & Specifications
Specifications
Reagents DNase Control Reagent
Product Type QC System with manual
Quantity 480 assays