Invitrogen™ DNaseAlert™ QC System DFS Item
Provides sensitive detection of DNases in a simple, easy-to-use fluorimetric assay with real time results
$1,016.00 - $1,016.00
Specifications
Quantity 480 assays
Reagents DNase Control Reagent
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
AM1970 View Documents Invitrogen™
LSAM1970
QC System Each for $1,016.00
Description
Description
Ambion™ DNaseAlert™ QC System is patent pending technology provides sensitive detection of DNases in simple, easy-to-use fluorimetric assay that delivers results in real time. Contains sufficient reagents for 5 × 96 high throughput assays.
  • Certify plastics, enzymes, solutions, and other biomaterials as in.DNase-free in. prior to DNase-sensitive applications such as PCR
  • Combine with RNaseAlert™ to quantitatively resolve both RNase and DNase activity simultaneously
  • Lypholized
  • DNaseAlert was designed to interface seamlessly with the RNaseAlert QC System (Cat. No. AM1966)
  • Permit two-color detection of both DNases and RNases in the same sample
  • Fluorescent reporter linked to DNaseAlert substrate is spectrally distinct from fluorescent tag on the RNaseAlert substrate
DNA and RNA Purification and Analysis, DNA Extraction, General gDNA Purification Reagents and Accessories, Genomic DNA Purification, PCR and Real-Time PCR, Real Time PCR (qPCR)
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
480 assays
DNase Control Reagent
For Research Use Only. Not for use in diagnostic procedures.