Please login to your online account to display your discounted pricing

Cloned AMV First-Strand cDNA Synthesis Kit DFS Item


Combines the high performance of Invitrogen Cloned AMV RT with the convenience of optimized, prequalified reagents

Manufacturer: life technologies  LS12328032

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12328032

  • / Each
 

Description & Specifications

Specifications

Includes 25μL cloned AMV RT (15U/μL), 500μL 5X cDNA synthesis buffer, 250μL 0.1M DTT, 100μL 10mM dNTP mix, 25μL RnaseOUT (40U/μL), 25μL oligo(dT)20 (50μM), 50μL random hexamers (50ng/μL), 1.25mL DEPC-treated water, product profile sheet
Temperature (Metric) Operating 45 to 55deg.C
Format Frozen
Temperature (English) Operating 113 to 131deg.F
For Use With (Application) First-Strand cDNA Synthesis, RT-PCR
Storage Requirements Store at -70deg.C
Packaging Type Tube
Test Sensitivity Low
Quantity 25 reactions (375 units)