Invitrogen™ Gateway™ LR Clonase™ Enzyme mix
Enzyme mix that offers high recombination efficiency
Manufacturer: Invitrogen™ 11791043
Description
Gateway LR Clonase enzyme mix contains proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes that catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate your expression clone.
- Specifically optimized for both single and multiple fragment cloning
- Provided in optimized single mix of enzyme and reaction buffer, ensuring enzyme stability and ease of use with few pippetting steps for both single and multiple fragment cloning
Cloning, Gateway Cloning, Gateway Recombination
Specifications
Dry Ice | |
Reaction Buffer | |
LR Clonase Enzyme Mix |
LR Clonase | |
100 Reactions | |
Clonase™, Gateway™ |
For Research Use Only. Not for use in diagnostic procedures.