Invitrogen™ Gateway™ BP Clonase™ II Enzyme mix
Catalyzes in vitro recombination of PCR products
Manufacturer: Invitrogen™ 11789020
Description
- Ideal for subcloning DNA segments from clones (containing attB sites) and donor vector (containing attP sites) to generate entry clones
- Gateway BP Clonase II contains enzymes and buffer in single mix to enable convenient ten-microliter reaction set up with fewer pipetting steps
Cloning, Gateway Cloning, Gateway Recombination
Specifications
Dry Ice | |
Enzyme Buffer | |
BP Clonase Enzyme Mix |
BP Clonase | |
20 Reactions | |
Clonase™, Gateway™ |
For Research Use Only. Not for use in diagnostic procedures.